2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
H-DL-Phg(4-CF₃)-OH, 4-(Trifluoromethyl)-DL-phenylglycine, 2-Amino-2-(4-(trifluoromethyl)phenyl)acetic acid
PC5857-5G
PC5857-1G
APOSPC5857-5GEA
14490
CZK
APOSPC5857-5G
APOSPC5857-1G
2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid
2-Amino-2-[4-(trifluoromethyl)phenyl]acetic acid
Vzorec:
C₉H₈F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00665333 CAS číslo: 142012-65-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...