Tisk…

(+/-)-α-Methoxy-α-trifluoromethylphenylacetyl chloride 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Methoxy-2-phenyl-3,3,3-trifluoropropanoyl chloride, (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

PC5847-25G PC5847-5G
APOSPC5847-25GEA 18650 CZK
APOSPC5847-25G APOSPC5847-5G
(+/-)-α-Methoxy-α-trifluoromethylphenylacetyl chloride 98%
(+/-)-alpha-Methoxy-alpha-trifluoromethylphenylacetyl chloride
Vzorec: C₁₀H₈ClF₃O₂
MW: 252,62 g/mol
Bod varu: 213…214 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078209
CAS číslo: 40793-68-8
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR