1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone 97+%
Dodavatel: Apollo Scientific
Synonyms:
5-Acetyl-2-(trifluoromethyl)pyridine
PC5837-1G
PC5837-250MG
PC5837-5G
APOSPC5837-1GEA
6120
CZK
APOSPC5837-1G
APOSPC5837-250MG
APOSPC5837-5G
1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone 97+%
1-(6-(Trifluoromethyl)pyridin-3-yl)ethanone
Vzorec:
C₈H₆F₃NO MW: 189.1346 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07375085 CAS číslo: 358780-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...