Tisk…

6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde

Dodavatel: Apollo Scientific
Varování

PC57784-1G PC57784-250MG
APOSPC57784-1GEA 8200 CZK
APOSPC57784-1G APOSPC57784-250MG
6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde
6-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde
Vzorec: C₁₀H₅F₃OS
MW: 230,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11840979
CAS číslo: 863118-49-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR