Tisk…

9-Fluorenylmethyl pentafluorophenyl carbonate 98%

Dodavatel: Apollo Scientific

Synonyms: Carbonic Acid 9-Fluorenylmethyl Pentafluorophenyl Ester, Fmoc-OPfp, 9-Fluorenylmethyl Pentafluorophenyl Carbonate

PC5773-1G PC5773-5G
APOSPC5773-1GEA 2940 CZK
APOSPC5773-1G APOSPC5773-5G
9-Fluorenylmethyl pentafluorophenyl carbonate 98%
9-Fluorenylmethyl pentafluorophenyl carbonate
Vzorec: C₂₁H₁₁F₅O₃
MW: 406.31 g/mol
Bod tání: 85…87 °C
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013261
CAS číslo: 88744-04-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR