Tisk…

α,4-Dibromo-3-fluorotoluene 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Bromo-3-fluorobenzyl bromide, 1-Bromo-4-(bromomethyl)-2-fluorobenzene

PC5721-5G PC5721-1G PC5721-25G
APOSPC5721-5GEA 744 CZK
APOSPC5721-5G APOSPC5721-1G APOSPC5721-25G
α,4-Dibromo-3-fluorotoluene 97%
α,4-Dibromo-3-fluorotoluene
Vzorec: C₇H₅Br₂F
MW: 267,92 g/mol
Bod varu: 112…114 °C/8mmHg
Bod tání: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD11847179
CAS číslo: 127425-73-4
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR