Tisk…

trans-4-Fluoro-L-proline

Dodavatel: Apollo Scientific
Varování

Synonyms: (2S,4R)-4-Fluoropyrrolidine-2-carboxylic Acid

PC5718-100MG PC5718-250MG
APOSPC5718-100MGEA 6650 CZK
APOSPC5718-100MG APOSPC5718-250MG
trans-4-Fluoro-L-proline
trans-4-Fluoro-L-proline
Vzorec: C₅H₈FNO₂
MW: 133.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683982
CAS číslo: 2507-61-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR