Tisk…

2-Fluoro-5-(trifluoromethoxy)phenol

Dodavatel: Apollo Scientific
Nebezpečí

PC5707-1G PC5707-5G PC5707-10G
APOSPC5707-1GEA 1330 CZK
APOSPC5707-1G APOSPC5707-5G APOSPC5707-10G
2-Fluoro-5-(trifluoromethoxy)phenol
2-Fluoro-5-(trifluoromethoxy)phenol
Vzorec: C₇H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04115893
CAS číslo: 886498-03-9
UN: 1760
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR