1H,1H-Perfluoropentylamine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
1H,1H-Nonafluoropentylamine
PC57021-25G
PC57021-5G
APOSPC57021-25GEA
20470
CZK
APOSPC57021-25G
APOSPC57021-5G
1H,1H-Perfluoropentylamine
1H,1H-Perfluoropentylamine
Vzorec:
C₅H₄F₉N MW: 249,08 g/mol Bod varu: 87 °C Hustota: 1,57 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS číslo:
355-27-1 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...