Tisk…

5-Bromo-1,3-difluoro-2-nitrobenzene 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,5-Difluoro-4-nitrobromobenzene

PC57010-25G PC57010-5G PC57010-10G
APOSPC57010-25GEA 4240 CZK
APOSPC57010-25G APOSPC57010-5G APOSPC57010-10G
5-Bromo-1,3-difluoro-2-nitrobenzene 98%
5-Bromo-1,3-difluoro-2-nitrobenzene
Vzorec: C₆H₂BrF₂NO₂
MW: 237,9864 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025434
CAS číslo: 147808-42-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR