7-Fluoro-1-benzothiophene-2-carbohydrazide
Dodavatel: Apollo Scientific
|
Varování
|
PC56975-1G
PC56975-5G
APOSPC56975-1GEA
622
CZK
APOSPC56975-1G
APOSPC56975-5G
7-Fluoro-1-benzothiophene-2-carbohydrazide
7-Fluoro-1-benzothiophene-2-carbohydrazide
Vzorec:
C₉H₇FN₂OS Storage Temperature: Ambient |
MDL Number:
MFCD11100241 CAS číslo: 1048913-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...