Tisk…

Cobalt(III) fluoride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Cobaltic fluoride, Cobalt trifluoride

PC5678-100G PC5678-25G PC5678-5G
APOSPC5678-100GEA 6450 CZK
APOSPC5678-100G APOSPC5678-25G APOSPC5678-5G
Cobalt(III) fluoride 99%
Cobalt(III) fluoride

Hard fluorinating agent. Replaces organic hydrogen with fluorine, J. Appl. Chem., 1952, 127.

Vzorec: CoF₃
MW: 115.93
Bod tání: 927
Hustota: 3.88 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010942
CAS číslo: 10026-18-3
EINECS: 233-062-4
UN: 3084
ADR: 8+5.1 II
Merck Index: 12,02491

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR