Tisk…

Potassium tetrakis(pentafluorophenyl)borate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Potassium tetrakis(perfluorophenyl)borate

PC56696-5G PC56696-1G
APOSPC56696-5GEA 2900 CZK
APOSPC56696-5G APOSPC56696-1G
Potassium tetrakis(pentafluorophenyl)borate 97%
Potassium tetrakis(pentafluorophenyl)borate
Vzorec: C₂₄BF₂₀K
MW: 718,14 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD06797410
CAS číslo: 89171-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR