Tisk…

3-(Trifluoromethoxy)phenylboronic acid pinacol ester

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Trifluoromethoxy)phenylboronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-[3-(trifluoromethoxy)phenyl]-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethoxy)benzene

PC5663-5G
APOSPC5663-5GEA 2070 CZK
APOSPC5663-5G
3-(Trifluoromethoxy)phenylboronic acid pinacol ester
3-(Trifluoromethoxy)phenylboronic acid pinacol ester
Vzorec: C₁₃H₁₆BF₃O₃
MW: 288,0706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863919
CAS číslo: 262376-31-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR