Tisk…

4-tert-Butyl-2,6-dimethylphenylsulphur trifluoride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-tert-Butyl-2,6-dimethylphenylsulphur trifluoride, FLUOLEAD(TM)

PC5654-1G PC5654-25G PC5654-5G
APOSPC5654-1GEA 622 CZK
APOSPC5654-1G APOSPC5654-25G APOSPC5654-5G
4-tert-Butyl-2,6-dimethylphenylsulphur trifluoride 95%
4-tert-Butyl-2,6-dimethylphenylsulphur trifluoride

Safer alternative to DAST.

Vzorec: C₁₂H₁₇F₃S
MW: 250.32 g/mol
Bod varu: 150 °C
Bod tání: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD11858620
CAS číslo: 947725-04-4
UN: 2923
ADR: 8+6.1 I

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR