Tisk…

3-Amino-4-methoxyphenylsulphonyl fluoride 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC5637-10G PC5637-50G PC5637-1G
APOSPC5637-10GEA 0 CZK
APOSPC5637-10G APOSPC5637-50G APOSPC5637-1G
3-Amino-4-methoxyphenylsulphonyl fluoride 97%
3-Amino-4-methoxyphenylsulphonyl fluoride
Vzorec: C₇H₈FNO₃S
Bod tání: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00007417
CAS číslo: 498-74-8
EINECS: 207-870-2
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR