Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%
Dodavatel: Apollo Scientific
|
Varování
|
PC56331-5G
PC56331-1G
APOSPC56331-5GEA
27280
CZK
APOSPC56331-5G
APOSPC56331-1G
Sodium tetrakis(4-fluorophenyl)borate dihydrate 99+%
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Vzorec:
C₂₄H₂₀BF₄NaO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00149598 CAS číslo: 207683-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...