Tisk…

4-Bromo-3-chloro-2-fluorobenzonitrile

Dodavatel: Apollo Scientific
Nebezpečí

PC56322-1G PC56322-250MG PC56322-5G
APOSPC56322-1GEA 3310 CZK
APOSPC56322-1G APOSPC56322-250MG APOSPC56322-5G
4-Bromo-3-chloro-2-fluorobenzonitrile
4-Bromo-3-chloro-2-fluorobenzonitrile
Vzorec: C₇H₂BrClFN
Storage Temperature: Ambient
MDL Number: MFCD11845913
CAS číslo: 1160574-68-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR