4-Amino-2,3,5,6-tetrafluoropyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2,3,5,6-Tetrafluoropyridin-4-amine
PC5620-1G
PC5620-5G
PC5620-10G
APOSPC5620-1GEA
622
CZK
APOSPC5620-1G
APOSPC5620-5G
APOSPC5620-10G
4-Amino-2,3,5,6-tetrafluoropyridine 98%
4-Amino-2,3,5,6-tetrafluoropyridine
Vzorec:
C₅H₂F₄N₂ MW: 166,08 g/mol Bod tání: 84…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010855 CAS číslo: 1682-20-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...