1,3-Dibromo-2,5-difluorobenzene 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC56083-1G
PC56083-5G
PC56083-25G
APOSPC56083-1GEA
622
CZK
APOSPC56083-1G
APOSPC56083-5G
APOSPC56083-25G
1,3-Dibromo-2,5-difluorobenzene 99%
1,3-Dibromo-2,5-difluorobenzene
Vzorec:
C₆H₂Br₂F₂ MW: 271.8900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09878120 CAS číslo: 128259-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...