Tisk…

1,4-Dibromo-2,3-difluorobenzene 95+%

Dodavatel: Apollo Scientific
Varování

PC56082-1G PC56082-5G PC56082-10G
APOSPC56082-1GEA 622 CZK
APOSPC56082-1G APOSPC56082-5G APOSPC56082-10G
1,4-Dibromo-2,3-difluorobenzene 95+%
1,4-Dibromo-2,3-difluorobenzene
Vzorec: C₆H₂Br₂F₂
Bod varu: 93…94 °C/50mmHg
Storage Temperature: Ambient
MDL Number: MFCD09835192
CAS číslo: 156682-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR