Tisk…

N-Methyl iodofluoroacetamide 95%

Dodavatel: Apollo Scientific

PC5599-1G PC5599-5G
APOSPC5599-1GEA 6050 CZK
APOSPC5599-1G APOSPC5599-5G
N-Methyl iodofluoroacetamide 95%
N-Methyl iodofluoroacetamide
Vzorec: C₃H₅FINO
Storage Temperature: Ambient
MDL Number: MFCD04038321
CAS číslo: 883499-47-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR