Tisk…

N-Methyl chlorofluoroacetamide 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-2-fluoro-N-methylacetamide

PC5596-5G PC5596-1G
APOSPC5596-5GEA 10820 CZK
APOSPC5596-5G APOSPC5596-1G
N-Methyl chlorofluoroacetamide 98%
N-Methyl chlorofluoroacetamide
Vzorec: C₃H₅ClFNO
MW: 125,53 g/mol
Bod varu: 80 °C/0,4mmHg
Storage Temperature: Ambient
MDL Number: MFCD04038320
CAS číslo: 53441-15-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR