Tisk…

N-Methyl bromofluoroacetamide 98%

Dodavatel: Apollo Scientific

PC5594-25G PC5594-5G PC5594-1G
APOSPC5594-25GEA 14090 CZK
APOSPC5594-25G APOSPC5594-5G APOSPC5594-1G
N-Methyl bromofluoroacetamide 98%
N-Methyl bromofluoroacetamide
Vzorec: C₃H₅BrFNO
Storage Temperature: Ambient
MDL Number: MFCD04038318
CAS číslo: 53441-14-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR