1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)-propan-2-ol 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC5582-1G
PC5582-5G
APOSPC5582-1GEA
2320
CZK
APOSPC5582-1G
APOSPC5582-5G
1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)-propan-2-ol 97%
1,1,1,3,3,3-Hexafluoro-2-(4-vinylphenyl)propan-2-ol
Vzorec:
C₁₁H₈F₆O MW: 270.18 g/mol Bod varu: 74…75 °C/3mmHg Storage Temperature: Ambient |
MDL Number:
MFCD04038310 CAS číslo: 2386-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...