1,1,2,2,3,3-Hexafluoropropane
Dodavatel: Apollo Scientific
|
Varování
|
PC5581-5G
PC5581-25G
APOSPC5581-5GEA
10490
CZK
APOSPC5581-5G
APOSPC5581-25G
1,1,2,2,3,3-Hexafluoropropane
1,1,2,2,3,3-Hexafluoropropane
Vzorec:
C₃H₂F₆ Bod varu: 5 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093293 CAS číslo: 27070-61-7 UN: 3163 ADR: 2.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...