Hexafluoroisopropyl crotonate 99%
Dodavatel: Apollo Scientific
PC5580-5G
PC5580-25G
APOSPC5580-5GEA
3740
CZK
APOSPC5580-5G
APOSPC5580-25G
Hexafluoroisopropyl crotonate 99%
Hexafluoroisopropyl crotonate
Vzorec:
C₇H₆F₆O₂ Bod varu: 62…63 °C/115mmHg Hustota: 1,3 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD02093322 CAS číslo: 135771-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...