Tisk…

Pentafluorobenzyl methacrylate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methacrylic acid Pentafluorobenzyl ester

PC5555-1G PC5555-25G PC5555-5G
APOSPC5555-1GEA 1210 CZK
APOSPC5555-1G APOSPC5555-25G APOSPC5555-5G
Pentafluorobenzyl methacrylate 97%
Pentafluorobenzyl methacrylate
Vzorec: C₁₁H₇F₅O₂
MW: 266.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042329
CAS číslo: 114859-23-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR