3-Fluoro-4-methoxybenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-Fluoro-4-methoxybenzene-1-sulphonyl chloride
PC5547-1G
PC5547-25G
PC5547-5G
APOSPC5547-1GEA
622
CZK
APOSPC5547-1G
APOSPC5547-25G
APOSPC5547-5G
3-Fluoro-4-methoxybenzenesulphonyl chloride
3-Fluoro-4-methoxybenzenesulphonyl chloride
Vzorec:
C₇H₆ClFO₃S MW: 224,64 g/mol Bod tání: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD02091380 CAS číslo: 67475-55-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...