Nonafluoro-3,6-dioxaheptanoic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC5519-25G
PC5519-5G
PC5519-1G
APOSPC5519-25GEA
8900
CZK
APOSPC5519-25G
APOSPC5519-5G
APOSPC5519-1G
Nonafluoro-3,6-dioxaheptanoic acid 98%
Nonafluoro-3,6-dioxaheptanoic acid
Vzorec:
C₅HF₉O₄ MW: 296,05 g/mol Bod varu: 145…146 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095398 CAS číslo: 151772-58-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...