Methyl-4,4,4-trifluorobutyrate
Dodavatel: Apollo Scientific
|
Varování
|
PC5516-1G
PC5516-5G
APOSPC5516-1GEA
1660
CZK
APOSPC5516-1G
APOSPC5516-5G
Methyl-4,4,4-trifluorobutyrate
Methyl-4,4,4-trifluorobutyrate
Vzorec:
C₅H₇F₃O₂ Bod varu: 113 °C Storage Temperature: Ambient |
CAS číslo:
2365-82-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...