5-Bromo-2,2-difluoro-1,3-benzodioxole 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
PC5489-100G
APOSPC5489-100GEA
1820
CZK
APOSPC5489-100G
5-Bromo-2,2-difluoro-1,3-benzodioxole 98%
5-Bromo-2,2-difluoro-1,3-benzodioxole
Vzorec:
C₇H₃BrF₂O₂ MW: 237 Bod varu: 78…79 °C/20mmHg Hustota: 1.74 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236212 CAS číslo: 33070-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...