Pentafluorobenzenesulphonyl chloride 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,3,4,5,6-Pentafluorobenzenesulphonyl chloride, Pentafluorobenzene-1-sulphonyl chloride
PC5480-100G
PC5480-1G
APOSPC5480-100GEA
0
CZK
APOSPC5480-100G
APOSPC5480-1G
Pentafluorobenzenesulphonyl chloride 99%
Pentafluorobenzenesulphonyl chloride
Vzorec:
C₆ClF₅O₂S MW: 266.58 Bod varu: 210…211 °C Hustota: 1.797 g/cm³ (25 °C) Bod vzplanutí: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007427 CAS číslo: 832-53-1 EINECS: 212-620-0 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...