Tisk…

Allyl-2-chloro-1,1,2-trifluoroethyl ether 97%

Dodavatel: Apollo Scientific

PC5460-25G PC5460-100G PC5460-5G
APOSPC5460-25GEA 5690 CZK
APOSPC5460-25G APOSPC5460-100G APOSPC5460-5G
Allyl-2-chloro-1,1,2-trifluoroethyl ether 97%
Allyl-2-chloro-1,1,2-trifluoroethyl ether
Vzorec: C₅H₆ClF₃O
MW: 174,55 g/mol
Bod varu: 109,2 °C/750mmHg
Hustota: 1,25 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039336
CAS číslo: 380-44-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR