Tisk…

Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane

Dodavatel: Apollo Scientific
Varování

PC5428-10G PC5428-5G
APOSPC5428-10GEA 992 CZK
APOSPC5428-10G APOSPC5428-5G
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Bis-2,2-[4-(4-nitrophenoxy)phenyl]hexafluoropropane
Vzorec: C₂₇H₁₆F₆N₂O₆
MW: 578,43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00317556
CAS číslo: 69563-87-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR