Tisk…

α-Chloro-2,5-difluorotoulene 97%

Dodavatel: Apollo Scientific
Nebezpečí

PC5408-100G PC5408-25G PC5408-5G
APOSPC5408-100GEA 13230 CZK
APOSPC5408-100G APOSPC5408-25G APOSPC5408-5G
α-Chloro-2,5-difluorotoulene 97%
α-Chloro-2,5-difluorotoulene
Vzorec: C₇H₅ClF₂
MW: 162,57 g/mol
Bod varu: 62…64 °C/14mmHg
Hustota: 1,3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01090989
CAS číslo: 495-07-8
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR