Tisk…

3-Fluoro-5-(trifluoromethyl)pyridine 97%

Dodavatel: Apollo Scientific

PC5384-10G PC5384-5G
APOSPC5384-10GEA 15340 CZK
APOSPC5384-10G APOSPC5384-5G
3-Fluoro-5-(trifluoromethyl)pyridine 97%
3-Fluoro-5-(trifluoromethyl)pyridine
Vzorec: C₆H₃F₄N
Storage Temperature: Ambient
MDL Number: MFCD11100535
CAS číslo: 1099597-96-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR