Tisk…

4-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)pentanoic acid 97%

Dodavatel: Apollo Scientific

PC5362-1G PC5362-500MG PC5362-5G
APOSPC5362-1GEA 6190 CZK
APOSPC5362-1G APOSPC5362-500MG APOSPC5362-5G
4-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)pentanoic acid 97%
4-Methyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)pentanoic acid
Vzorec: C₁₃H₁₆F₃NO₄S
Bod tání: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00173033
CAS číslo: 251097-65-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR