Tisk…

2-Bromo-6-chloro-4-(trifluoromethoxy)aniline

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-6-chloro-4-trifluoromethoxyaniline

PC53547-1G PC53547-5G PC53547-25G
APOSPC53547-1GEA 2690 CZK
APOSPC53547-1G APOSPC53547-5G APOSPC53547-25G
2-Bromo-6-chloro-4-(trifluoromethoxy)aniline
2-Bromo-6-chloro-4-(trifluoromethoxy)aniline
Vzorec: C₇H₄BrClF₃NO
MW: 290,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618925
CAS číslo: 1228108-74-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR