Tisk…

Perfluorotoluene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Octafluorotoluene, Pentafluoro(trifluoromethyl)benzene, 1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzene, α,α,α,2,3,4,5,6-Octafluorotoluene

PC5354-100G PC5354-25G
APOSPC5354-100GEA 5460 CZK
APOSPC5354-100G APOSPC5354-25G
Perfluorotoluene 98%
Perfluorotoluene

In base, hydroxyl groups react at the 4-position to yield heptafluoro-p-tolyl ethers, which can be used to protect estradiols.

Vzorec: C₇F₈
MW: 236,06 g/mol
Bod varu: 103…105 °C
Bod tání: –65,6 °C
Hustota: 1,666 g/cm³ (25 °C)
Bod vzplanutí: 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000375
CAS číslo: 434-64-0
EINECS: 207-104-7
UN: 1992
ADR: 3+6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR