Tisk…

1H,1H,5H-Octafluoropentyl methacrylate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methacrylic Acid 1H,1H,5H-Perfluoropentyl Ester, Methacrylic Acid 1H,1H,5H-Octafluoropentyl Ester, 1H,1H,5H-Perfluoropentyl Methacrylate, 2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate

PC5353-100G PC5353-25G PC5353-5G
APOSPC5353-100GEA 3540 CZK
APOSPC5353-100G APOSPC5353-25G APOSPC5353-5G
1H,1H,5H-Octafluoropentyl methacrylate 98%
1H,1H,5H-Octafluoropentyl methacrylate
Vzorec: C₉H₈F₈O₂
MW: 300.15 g/mol
Bod varu: 178…179 °C
Hustota: 1.432 g/cm³ (25 °C)
Bod vzplanutí: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00039278
CAS číslo: 355-93-1
EINECS: 206-596-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR