4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC53378-10G
PC53378-1G
PC53378-5G
APOSPC53378-10GEA
4240
CZK
APOSPC53378-10G
APOSPC53378-1G
APOSPC53378-5G
4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde
Vzorec:
C₇H₃BrFNO₃ MW: 248,0060 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10758067 CAS číslo: 679839-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...