Tisk…

1-Bromo-2-chloro-4-(trifluoromethoxy)benzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-4-(Trifluoromethoxy)bromobenzene

PC53328-1G PC53328-25G PC53328-5G PC53328-100G
APOSPC53328-1GEA 582 CZK
APOSPC53328-1G APOSPC53328-25G APOSPC53328-5G APOSPC53328-100G
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene
1-Bromo-2-chloro-4-(trifluoromethoxy)benzene
Vzorec: C₇H₃BrClF₃O
MW: 275,4500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111892
CAS číslo: 892845-59-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR