4-Bromo-3,5-difluorobenzonitrile 97%
Dodavatel: Apollo Scientific
PC53312-1G
PC53312-5G
APOSPC53312-1GEA
870
CZK
APOSPC53312-1G
APOSPC53312-5G
4-Bromo-3,5-difluorobenzonitrile 97%
4-Bromo-3,5-difluorobenzonitrile
Vzorec:
C₇H₂BrF₂N MW: 217.9983 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11845964 CAS číslo: 123688-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...