Tisk…

3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Fluoro-1H-pyrazolo[3,4-b]pyridin-3-amine

PC53221-1G PC53221-250MG
APOSPC53221-1GEA 8240 CZK
APOSPC53221-1G APOSPC53221-250MG
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₆H₅FN₄
MW: 152,13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15526909
CAS číslo: 1034667-22-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR