Tisk…

Octafluoroadipic acid 97%

Dodavatel: Apollo Scientific
Varování

PC5320-5G PC5320-250MG PC5320-1G
APOSPC5320-5GEA 2280 CZK
APOSPC5320-5G APOSPC5320-250MG APOSPC5320-1G
Octafluoroadipic acid 97%
Octafluoroadipic acid
Vzorec: C₆H₂F₈O₄
Bod varu: 165 °C/1mmHg
Bod tání: 132…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00066405
CAS číslo: 336-08-3
EINECS: 206-407-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR