2-[4-(2-Fluoro-4-nitrophenyl)piperazin-1-yl]ethanol
Dodavatel: Apollo Scientific
Synonyms:
1-(2-Hydroxyethyl)-4-(2-fluoro-4-nitrophenyl)piperazine
PC5317-1G
PC5317-5G
APOSPC5317-1GEA
4010
CZK
APOSPC5317-1G
APOSPC5317-5G
2-[4-(2-Fluoro-4-nitrophenyl)piperazin-1-yl]ethanol
2-[4-(2-Fluoro-4-nitrophenyl)piperazin-1-yl]ethanol
Vzorec:
C₁₂H₁₆FN₃O₃ MW: 269,28 g/mol Bod tání: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD08692441 CAS číslo: 942474-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...