(Bromodifluoromethyl)trimethylsilane 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Bromodifluoro(trimethylsilyl)methane
PC53035-5G
PC53035-25G
PC53035-1G
APOSPC53035-5GEA
3110
CZK
APOSPC53035-5G
APOSPC53035-25G
APOSPC53035-1G
(Bromodifluoromethyl)trimethylsilane 97%
(Bromodifluoromethyl)trimethylsilane
Vzorec:
C₄H₉BrF₂Si Bod varu: 112 °C Hustota: 1,41 g/cm³ (25 °C) Bod vzplanutí: 45 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD18641931 CAS číslo: 115262-01-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...