2-Fluoro-4-(trifluoromethoxy)benzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Fluoro-4-(trifluoromethoxy)benzoic acid
PC53010-5G
PC53010-1G
APOSPC53010-5GEA
10350
CZK
APOSPC53010-5G
APOSPC53010-1G
2-Fluoro-4-(trifluoromethoxy)benzoic acid 95%
2-Fluoro-4-(trifluoromethoxy)benzoic acid
Vzorec:
C₈H₄F₄O₃ MW: 224.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16652439 CAS číslo: 1073477-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...