Tisk…

4-(4-Fluorophenoxy)piperidine hydrochloride 97%

Dodavatel: Apollo Scientific

PC530028-1G PC530028-5G
APOSPC530028-1GEA 8900 CZK
APOSPC530028-1G APOSPC530028-5G
4-(4-Fluorophenoxy)piperidine hydrochloride 97%
4-(4-Fluorophenoxy)piperidine hydrochloride
Vzorec: C₁₁H₁₅ClFNO
MW: 231,690 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115015
CAS číslo: 3202-34-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR